missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3-Pentafluorobutane, 99.5+%

Product Code. 15426597
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Product Code. 15426597

Brand: Thermo Scientific Alfa Aesar H33737.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 406-58-6 | C4H5F5 | 148.076 g/mol

Used as foam blowing agent in the production of closed cell rigid insulating foams. It is also used as degreasing solvent in semi-closed and closed degreasing systems. It finds also its use as heat exchange fluid in closed heat transfer systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as foam blowing agent in the production of closed cell rigid insulating foams. It is also used as degreasing solvent in semi-closed and closed degreasing systems. It finds also its use as heat exchange fluid in closed heat transfer systems.

Solubility
Soluble in water (1.7g/L at 21.2C). Miscible with most organic solvents.

Notes
Store in cool location. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406-58-6
Molecular Formula C4H5F5
Molecular Weight (g/mol) 148.076
MDL Number MFCD03094115
InChI Key WZLFPVPRZGTCKP-UHFFFAOYSA-N
PubChem CID 67884
IUPAC Name 1,1,1,3,3-pentafluorobutane
SMILES CC(CC(F)(F)F)(F)F

Specifications

Density 1.27
Boiling Point 40°C
Flash Point −20°C (−4°F)
Linear Formula CH3CF2CH2CF3
Quantity 1000 g
UN Number UN1993
Synonym hfc-365mfc, butane, 1,1,1,3,3-pentafluoro, butane,1,1,1,3,3-pentafluoro, solkane 365 mfc, 2,2,4,4,4-pentafluorobutane, 1h,1h,1h,3h,3h-perfluorobutane
Solubility Information Soluble in water (1.7g/L at 21.2C). Miscible with most organic solvents.
Formula Weight 148.08
Percent Purity ≥99.5%
Chemical Name or Material 1,1,1,3,3-Pentafluorobutane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.