missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, 98%

Product Code. 10489220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
10mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10489220 500 mL Glass bottle 500mL
10479030 10 mL Glass bottle 10mL
10568470 100 mL Glass bottle 100mL
11985791 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10489220 Supplier Thermo Scientific Acros Supplier No. 120585000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 999-97-3 | C6H19NSi2 | 161.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.4
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Melting Point -78.0°C
Color Colorless
Density 0.7600g/mL
Boiling Point 125.0°C
Flash Point 20°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiNHSi(CH3)3
Refractive Index 1.4060 to 1.4090
Quantity 500 mL
Merck Index 15,4725
Specific Gravity 0.76
Solubility Information Solubility in water: reacts. Other solubilities: reacts with methanol, soluble in acetone, benzene, ethyl ether, pentane, and perchloroethylene
Viscosity 0.9 mm2/s (25°C)
Formula Weight 161.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexamethyldisilazane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.