missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™

Product Code. p-4229236 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
This item is not returnable. View return policy

Product Code. 10430398

Brand: Thermo Scientific Acros 430851000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 999-97-3 | C6H19NSi2 | 161.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.4
MDL Number MFCD00008259
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -78.0°C
Density 0.7600g/mL
Boiling Point 125.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)3SiNHSi(CH3)3
Refractive Index 1.4060 to 1.4090
Quantity 100 mL
Merck Index 15,4725
Specific Gravity 0.76
Solubility Information Solubility in water: reacts. Other solubilities: reacts with methanol, soluble in acetone, benzene, ethyl ether, pentane, and perchloroethylene
Viscosity 0.9 mm2/s (25°C)
Formula Weight 161.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexamethyldisilazane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.