missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-methoxyphenol

Product Code. 11318635
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318635 5 g 5g
11328635 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318635 Supplier Thermo Scientific Alfa Aesar Supplier No. L12128.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in agrochemical, pharmaceutical and dyestuff field. | CAS: 2160-89-6 | C6H4F6O2 | 222.09 g/mol

1,1,1,3,3,3-Hexafluoroisopropyl acrylate is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2160-89-6
Molecular Formula C6H4F6O2
Molecular Weight (g/mol) 222.09
MDL Number MFCD00040104
InChI Key MNSWITGNWZSAMC-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoroisopropyl acrylate, hexafluoroisopropyl acrylate, 1,1,1,3,3,3-hexafluoropropan-2-yl acrylate, 2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, hexafluoro-2-propyl acrylate, mnswitgnwzsamc-uhfffaoysa, 1h-1-trifluoromethyl trifluoroethyl acrylate
PubChem CID 75096
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
SMILES FC(F)(F)C(OC(=O)C=C)C(F)(F)F

Specifications

Color Colorless
Boiling Point 81°C to 82°C
Flash Point 17°C
Packaging Glass bottle
Linear Formula H2C=CHCO2CH(CF3)2
Quantity 5 g
UN Number UN3272
Specific Gravity 1.37
Formula Weight 222.09
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl acrylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.