missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroacetone sesquihydrate, 98%

Product Code. 10591491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591491 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10591491 Supplier Thermo Scientific Acros Supplier No. 269970250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13098-39-0 | 1·5 H2O | 193.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13098-39-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C to 21°C
Color Amber or Yellow
Density 1.6900g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Plastic bottle
Molecular Formula 1·5 H2O
Refractive Index 1.3060 to 1.311
MDL Number MFCD06800792
Quantity 25 g
Specific Gravity 1.69
Synonym hexafluoroacetone sesquihydrate, unii-cu1noz8ps6, ccris 4653, acetone, hexafluoro-, sesquihydrate, hexafluoro-2-propanone sesquihydrate, cu1noz8ps6, 2-propanone, hexafluoro-, sesquihydrate, 1,1,1,3,3,3-hexafluoroacetone sesquihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, hydrate 2:3, hexafluoroacetone 1.5-hydrate
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
InChI Key VAIZVCMDJPBJCM-UHFFFAOYSA-N
SMILES C(=O)(C(F)(F)F)C(F)(F)F.C(=O)(C(F)(F)F)C(F)(F)F.O.O.O
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
Molecular Weight (g/mol) 193.05
PubChem CID 3032602
Viscosity 0.67 mPa.s (38°C)
Formula Weight 193.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone sesquihydrate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoroacetone sesquihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.