missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, TRC

Product Code. 17992775
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
Unit Size:
25g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992775 25 g 25g
17952775 500 g 500g
17912345 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992775 Supplier Toronto Research Chemicals Supplier No. TRCH29388025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3 H2 F6 O
Molecular Weight (g/mol) 168.04
Synonym 2-Propanol, 1,1,1,3,3,3-hexafluoro-, 1,1,1,3,3,3-Hexafluoro-2-propanol, 1,1,1,3,3,3-Hexafluoro-2-hydroxypropane, 1,1,1,3,3,3-Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, 1,1,1,3,3,3-Hexafluoropropane-2-ol, 1,1,1,3,3,3-Hexafluoropropanol, 2,2,2-Trifluoro-1-(trifluoromethyl)ethanol, Bis(trifluoromethyl)methanol, HFIP, Hexafluoroisopropanol, Hexafluoroisopropyl alcohol, NSC 96336, 2H-Perfluoro-2-propanol
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES OC(C(F)(F)F)C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H
Formula Weight 168.001
Chemical Name or Material 1,1,1,3,3,3-hexafluoro-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.