missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, LC-MS Grade, Honeywell Fluka™

Product Code. p-7120006 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 mL
10 x 1 mL
Packaging:
Ampoule
Glass Bottle
Unit Size:
50mL
Pack of 10
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Product Code. 15688400

Brand: Honeywell-Fluka 4206010X1ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point -4°C
Color Colorless
Density 1.596g/mL at 25°C
Boiling Point 59°C
Packaging Ampoule
Linear Formula (CF3)2CHOH
Refractive Index n20/D 1.275
Quantity 10 x 1 mL
UN Number UN3265
Beilstein 1841007
Residue after Evaporation 0 to 0.0005%
Formula Weight 168.04g/mol
Percent Purity ≥99.8% (GC)
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,1,1,3,3,3-Hexafluoropropan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.