missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic)

Product Code. p-7051944
Click to view available options
Quantity:
1 g
5 x 1 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11314338

Brand: Thermo Scientific Alfa Aesar 042291.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 38701-74-5 | C3H2F6O | 170.05 g/mol

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2} is a stable isotope used for biochemical research. It is also used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38701-74-5
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 170.05
MDL Number MFCD00037559
InChI Key BYEAHWXPCBROCE-AWPANEGFSA-N
PubChem CID 2724852
SMILES [2H]OC([2H])(C(F)(F)F)C(F)(F)F

Specifications

Physical Form Liquid
Density 1.60 g/mL
Packaging Prescored Ampule
Sensitivity Moisture
Quantity 1 g
Melting Point −4°C
Boiling Point 59°C
UN Number UN1760
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 1,1,1,3,3,3-hexafluoro 2-2h propan-2-2h ol, hexafluoro 2 h propan-2-2 h ol, hexafluoro 2h propan-2-2h ol, 1,1,1,3,3,3-hexafluoro-2-propanol-d2 hfip-d2, 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 99 atom % d
Solubility Information Fully miscible in water.
IUPAC Name 2-deuterio-2-deuteriooxy-1,1,1,3,3,3-hexafluoropropane
Formula Weight 170.05
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol-d2, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.