missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol, 98+%

Product Code. 11479307
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11479307

Brand: Thermo Scientific Alfa Aesar B21366.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 718-64-9 | C9H6F6O | 244.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 718-64-9
Molecular Formula C9H6F6O
Molecular Weight (g/mol) 244.136
MDL Number MFCD00040983
InChI Key IZPIZCAYJQCTNG-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-phenyl-2-propanol, hexafluoro-2-hydroxyisopropyl benzene, bis trifluoromethyl phenylcarbinol, 2,2,2,2',2',2'-hexafluorocumyl alcohol, alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2-phenylisopropanol, benzenemethanol, .alpha.,.alpha.-bis trifluoromethyl, hexafluoro-2-phenyl isopropanol, hexafluoro-2-phenyl-2-propanol, benzenemethanol, alpha,alpha-bis trifluoromethyl
PubChem CID 69748
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol
SMILES C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Density 1.45
Boiling Point 159°C to 160°C
Flash Point 60°C (140°F)
Refractive Index 1.415
Quantity 25 g
UN Number UN1987
Beilstein 2053547
Formula Weight 244.14
Percent Purity ≥98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.