missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2,2-Pentafluoro-3-iodopropane, 96%

Product Code. 11307516
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307516 1 g 1g
11317516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307516 Supplier Thermo Scientific Alfa Aesar Supplier No. L11047.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 354-69-8 | C3H2F5I | 259.945 g/mol

1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-69-8
Molecular Formula C3H2F5I
Molecular Weight (g/mol) 259.945
MDL Number MFCD00039405
InChI Key HENALDZJQYAUBN-UHFFFAOYSA-N
Synonym pentafluoropropyl iodide, 2,2,3,3,3-pentafluoropropyl iodide, propane, 1,1,1,2,2-pentafluoro-3-iodo, 1h,1h-pentafluoropropyl iodide, 1-iodo-2,2,3,3,3-pentafluoropropane, acmc-1cmxs, cf3cf2ch2i, 2,3,3,3-pentafluoropropyl iodide, 3-iodo-1,1,1,2,2-pentafluoropropane, propane,1,1,2,2-pentafluoro-3-iodo
PubChem CID 67721
IUPAC Name 1,1,1,2,2-pentafluoro-3-iodopropane
SMILES C(C(C(F)(F)F)(F)F)I

Specifications

Boiling Point 70°C to 71°C
Refractive Index 1.373
Quantity 1 g
Beilstein 1700278
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 259.95
Percent Purity 96%
Chemical Name or Material 1,1,1,2,2-Pentafluoro-3-iodopropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.