Learn More
Greiner Bio-One 0.7mL Polypropylene Microcentrifuge Tubes
Product Code. 13077793
Click to view available options
Color:
Blue
Unit Size:
Pack of 500
Product Code. 13077793
Brand: Greiner Bio-One 742270
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For single use only. Greiner Bio-One Cell 0.7mL Polypropylene Microcentrifuge Tube is free of detectable DNase, RNase, human DNA.
  • Material: Polypropylene
  • For single use only
  • With attached cap
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Non-Pyrogenic
  • High chemical and temperature resistance
  • Suitable for Roche centrifu
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polypropylene
Volume (Metric) 0.7mL
Sterility Non-sterile