missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17C Antibody (177114), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1234M
This item is not returnable.
View return policy
Description
IL-17C Monoclonal antibody specifically detects IL-17C in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF
Specifications
| IL-17C | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 177114 | |
| Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
| E. coli-derived recombinant human IL-17C, His19-Val197, Accession # Q9P0M4.1 | |
| 0.5 mL | |
| Cytokine Research | |
| 27189 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction