missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Monkey IgG Heavy Chain Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60692F
This item is not returnable.
View return policy
Description
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG Heavy Chain | |
| Polyclonal | |
| PBS | |
| Goat | |
| Multi-step | |
| RUO | |
| Monkey | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| FITC | |
| Immunoglobulin G, ImmunoglobulinG | |
| Goat anti-Monkey IgG (gamma chain) Secondary Antibody was produced by repeated immunization with monkey IgG gamma heavy chain in goat. | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction