missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Epitope Tag Horseradish Peroxidase-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems HAM85291
This item is not returnable.
View return policy
Description
DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Multi-Species samples. It is validated for Western Blot.
Specifications
| DYKDDDDK Epitope Tag | |
| Monoclonal | |
| HRP | |
| Supplied as a 0.2 μm filtered solution in PBS with No Preservative | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects N-terminal and C-terminal DYKDDDDK (FLAG™) epitope tagged proteins in Western blot. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
| Western Blot | |
| 1042E | |
| Western Blot 1:1000-1:3000 dilution | |
| DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
| DYKDDDDK epitope tag | |
| 0.5 mL | |
| Primary | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction