missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin AP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems APM7520
This item is not returnable.
View return policy
Description
Digoxigenin Monoclonal specifically detects Digoxigenin in Multi-Species samples. It is validated for Immunohistochemistry.
Specifications
| Digoxigenin | |
| Monoclonal | |
| Alkaline Phosphatase (AP) | |
| Supplied as a 10X solution of antibody in 0.5 mL 0.2 M Tris buffer, pH7.3, with 0.1 mM ZnCl2, 0.4 mM MgCl2 and 0.09% sodium azide. with 0.09% Sodium Azide | |
| Mouse | |
| RUO | |
| Multi-species | |
| IgG2a |
| Immunohistochemistry | |
| 611621 | |
| Immunohistochemistry | |
| DIG | |
| 0.5 mL | |
| Primary | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction