Learn More
ChemR23/CMKLR1 Antibody (84939), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB362PECY55
Product Code. 30038425
  • £928.00 / 0.50mL
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ChemR23/CMKLR1
Monoclonal
PE-Cyanine5.5
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
84939
Flow Cytometry
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
NS0 mouse myeloma cell line transfected with human ChemR23, Met1-Leu371, Accession # NP_004063
0.5 mL
GPCR, Immunology
1240
Store at 4C in the dark. Do not freeze.
IgG3