missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zonisamide-d4, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Zonisamide-D4 |
| CAS | 1020720-04-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 2H4 H4 N2 O3 S |
| Quantity | 0.5 mg |
| Synonym | Zonisamide-D4 |
| InChI Formula | InChI=1S/C8H8N2O3S/c9-14(11,12)5-7-6-3-1-2-4-8(6)13-10-7/h1-4H,5H2,(H2,9,11,12)/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c([2H])c2c(CS(=O)(=O)N)noc2c1[2H] |
| IUPAC Name | (4,5,6,7-tetradeuterio-1,2-benzoxazol-3-yl)methanesulfonamide |
| Molecular Weight (g/mol) | 216.25 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction