missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Venlafaxine-d6 Hydrochloride (N,N-dimethyl-d6), TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Venlafaxine-D6 Hydrochloride |
| CAS | 1062606-12-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 2H6 H21 N O2 . Cl H |
| Quantity | 0.5 mg |
| Synonym | Cyclohexanol','1-[2-[di(methyl-d3)amino]-1-(4-methoxyphenyl)ethyl]-','hydrochloride (1:1), Venlafaxine-D6 Hydrochloride |
| InChI Formula | InChI=1S/C17H27NO2.ClH/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14;/h7-10,16,19H,4-6,11-13H2,1-3H3;1H/i1D3,2D3; |
| SMILES | Cl.[2H]C([2H])([2H])N(CC(c1ccc(OC)cc1)C2(O)CCCCC2)C([2H])([2H])[2H] |
| IUPAC Name | 1-[2-[bis(trideuteriomethyl)amino]-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride |
| Molecular Weight (g/mol) | 319.9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction