missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticagrelor-d7, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Ticagrelor-d7 |
| CAS | 1265911-55-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 2H7 H21 F2 N6 O4 S |
| Quantity | 0.5 mg |
| Synonym | (1S,2S,3R,5S)-3-[7-[[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino]-5-(propyl-1,1,2,2,3,3,3-d7-thio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)-1,2-cyclopentanediol, Ticagrelor-d7 |
| InChI Formula | InChI=1S/C23H28F2N6O4S/c1-2-7-36-23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(35-6-5-32)20(34)19(16)33/h3-4,8,12,15-17,19-20,32-34H,2,5-7,9-10H2,1H3,(H,26,27,28)/t12-,15+,16+,17-,19-,20+/m0/s1/i1D3,2D2,7D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])Sc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCO)[C@@H](O)[C@H]5O)c4n1 |
| IUPAC Name | (1S,2S,3R,5S)-3-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-(1,1,2,2,3,3,3-heptadeuteriopropylsulfanyl)triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)cyclopentane-1,2-diol |
| Molecular Weight (g/mol) | 529.61 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction