missing translation for 'onlineSavingsMsg'
Learn More

Metolachlor-d11, TRC

Product Code. 30179533
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.50mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179533 0.5 mg 0.50mg
30179084 1 mg 1mg
30179267 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179533 Supplier Toronto Research Chemicals Supplier No. TRCM3387030.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1632119-30-2
Molecular Formula C15H11D11ClNO2
Molecular Weight (g/mol) 294.86
Synonym 2-Chloro-N-(1-methoxypropan-2-yl)-N-[3,4,5-trideuterio-2-(1,1,2,2,2-pentadeuterioethyl)-6-(trideuteriomethyl)phenyl]acetamide;
IUPAC Name 2-chloro-N-(1-methoxypropan-2-yl)-N-[3,4,5-trideuterio-2-(1,1,2,2,2-pentadeuterioethyl)-6-(trideuteriomethyl)phenyl]acetamide
SMILES [2H]c1c([2H])c(c(N(C(C)COC)C(=O)CCl)c(c1[2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3/i1D3,2D3,5D2,6D,7D,8D
Formula Weight 294.203
Chemical Name or Material Metolachlor-d11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.