missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgD Heavy Chain Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68532
This item is not returnable.
View return policy
Description
IgD Heavy Chain Polyclonal antibody specifically detects IgD Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Specifications
| IgD Heavy Chain | |
| Polyclonal | |
| FITC | |
| PBS | |
| Goat | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
| 0.5 mg/mL | |
| Flow Cytometry Neat - 1:10, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence Neat - 1:10 | |
| Constant Region of Heavy Chain of IgD, Ig Delta Chain C Region, Ig Delta Chain C Region NIG-65, Ig Delta Chain C Region WAH, Immunoglobulin Delta, immunoglobulin heavy constant delta | |
| Human IgD | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction