missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ 0.45μm Pore Cellulose Nitrate Membrane

Product Code. 10281601 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
47 mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10281601 47 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10281601 Supplier Sartorius™ Supplier No. 1380647ACR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Filter identification and lot number are printed on each envelope, ensuring GLP conformance

47mm membranes, with 3.1mm x 3.1mm grids, assure optimal retention and colony growth. Select a color to give the best contrast to your colonies. Filter identification and lot number are printed on each envelope, ensuring GLP conformance. Individually packed.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color Green/Green Grid
Filter Type Membrane Filter
Material Cellulose Nitrate
Pore Size 0.45 μm
Flow Rate 70mL/min.
For Use With (Application) Microbiological testing of water, pharmaceutical products, foods, beverages and similar products
No. per Pack 1000/Pack
Thickness (Metric) 0.115 to 0.145 mm
Type Membrane Filter
Diameter (Metric) 47 mm
Show More Show Less
compliance-icons
Product Identifier
  • FILTERS, MEMBRANE DISC STERILE GREEN WITH D=47mm
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.