Learn More
Thermo Scientific™ Tubes, strips of 8, blue
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
Brand: Thermo Scientific™ AB0264B
103.04 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 03
Additional Details : Weight : 0.42000kg
Description
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
- Compatible with 0.2 mL thermal cycler blocks
- 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Specifications
Certified DNA/RNase Free | |
0.2 mL | |
250 Strips | |
Non-sterile | |
Standard | |
250 Strip Tubes |
Blue | |
0.25 mL | |
Thermal Cyclers | |
Polypropylene | |
Not Included | |
8-Strip Tube |