missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes, strips of 8, green

Product Code. 10113703
Click to view available options
Quantity:
250 strips
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 10113703

Brand: Thermo Scientific™ AB0264G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.

Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.

Highlights

  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Green
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 250 strips
For Use With (Equipment) Thermal Cyclers
Sterility Non-sterile
Material Polypropylene
Reaction Speed Standard
Closure Type Not Included
Format 250 Strip Tubes
Product Type 8-strip Tube
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.