missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes (white) and Ultra Clear Caps, strips of 8

Product Code. 11504804
Change view
Click to view available options
Quantity:
10 Packs of 12 Strips
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11504804 10 Packs of 12 Strips Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11504804 Supplier Thermo Scientific™ Supplier No. AB1191

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.

Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.

Highlights

  • Compatible with 0.2 mL thermal cycler blocks
  • 8 tubes per strip
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color White, Ultra Clear
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 10 Packs of 12 Strips
For Use With (Equipment) Thermal Cyclers or Real-Time PCR Systems
Material Polypropylene
Reaction Speed Standard
Closure Type Flat Cap
No. of Wells 96
Format 120 Strip Tubes and Caps
Product Type 8-strip Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.