missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tubes and Domed Caps, strips of 8, assorted colors
Description
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
- Compatible with 0.2 mL thermal cycler blocks 8 tubes per strip 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips
- Ultra-Clear cap options available for use in qPCR assays
- PCR
- qPRC
- Real-Time PCR
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Color | Assorted |
| Volume (Metric) Working | 0.2 mL |
| Volume (Metric) | 0.25 mL |
| Quantity | 250 Strips |
| For Use With (Equipment) | Thermal Cyclers |
| Material | Polypropylene |
| Reaction Speed | Standard |
| Closure Type | Domed Cap |
| No. of Wells | 96 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?