missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 0.2 mL Polypropylene PCR Tube Strips

Product Code. 16272591 Shop All Axygen Scientific Products
Change view
Click to view available options
Closure Type:
Domed Cap
Flat Cap
Unit Size:
Pack of 1260
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type unitSize
16272591 Domed Cap Pack of 1260
16282591 Flat Cap Pack of 1260
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16272591 Supplier Axygen™ Supplier No. PCR0208ADC
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2 mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to prevent sample loss or without caps.

Axygen™ 0.2 mL thin-wall 8 strip PCR tubes, available in an assortment of colors are designed to fit a wide variety of thermal cyclers. Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.

  • RNase-/DNase-free
  • Nonpyrogenic
  • non-sterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Caps are included
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Volume (Metric) 0.2 mL
Quantity 1260 Strips/Case (8 Tubes/Strip)
Sterility Non-sterile
Closure Type Domed Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.