missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF687 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41175AF647
This item is not returnable.
View return policy
Description
ZNF687 Polyclonal antibody specifically detects ZNF687 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ZNF687 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 57592 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 647 | |
| KIAA1441DKFZp781I1719, zinc finger protein 687 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZNF687. The immunogen is located within amino acids 1160 - 1210 of ZNF687. Amino Acid Squence: DGEEEAPPSRSDPDGGD | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction