missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX3 Antibody (PCRP-ZHX3-1D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14225CL1
This item is not returnable.
View return policy
Description
ZHX3 Monoclonal antibody specifically detects ZHX3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Protein Array
Specifications
| ZHX3 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
| Recombinant full-length human ZHX3 protein (Uniprot: Q9H4I2) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Protein Array | |
| PCRP-ZHX3-1D11 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 23051 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction