missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-C Antibody (107/F10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18626UV
This item is not returnable.
View return policy
Description
VEGF-C Monoclonal antibody specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| VEGF-C | |
| Monoclonal | |
| DyLight 350 | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| Recombinant human VEGF-C | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
| 7424 | |
| Store at 4°C in the dark. | |
| IgG2 α |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 107/F10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction