missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VBP1 Antibody (OTI2E6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74836CL1
This item is not returnable.
View return policy
Description
VBP1 Monoclonal antibody specifically detects VBP1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| VBP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| HIBBJ46, PFD3, PFDN3VHL-binding protein 1, prefoldin 3, prefoldin subunit 3, VBP-1, VHL binding protein-1, von Hippel-Lindau binding protein 1, Von Hippel-Lindau-binding protein 1 | |
| Full length human recombinant protein of human VBP1(NP_003363) produced in HEK293T cell. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7411 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI2E6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction