missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vav-1 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18452JF669
This item is not returnable.
View return policy
Description
Vav-1 Polyclonal antibody specifically detects Vav-1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Vav-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 669 | |
| oncogene vav, proto-oncogene vav, vav 1 guanine nucleotide exchange factor, vav proto-oncogene, VAVvav 1 oncogene | |
| Recognizes region between residues Between Between 175 and 225.. | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 7409 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction