missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tropomyosin-1 Antibody (TPM1/4510) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27021APC
This item is not returnable.
View return policy
Description
Tropomyosin-1 Monoclonal antibody specifically detects Tropomyosin-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Tropomyosin-1 | |
| Monoclonal | |
| APC | |
| alpha tropomyosin, alpha-tropomyosin, C15orf13, cardiomyopathy, hypertrophic 3, chromosome 15 open reading frame 13, CMD1Y, CMH3, HTM-alpha, sarcomeric tropomyosin kappa, TMSA, tropomyosin 1 (alpha), tropomyosin 1 (alpha) isoform 1, tropomyosin 1 (alpha) isoform 2, tropomyosin 1 (alpha) isoform 3, tropomyosin 1 (alpha) isoform 4, tropomyosin 1 (alpha) isoform 5, tropomyosin 1 (alpha) isoform 6, tropomyosin 1 (alpha) isoform 7, tropomyosin alpha-1 chain, Tropomyosin-1 | |
| Recombinant full-length Tropomyosin-1 protein | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 7168 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| TPM1/4510 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction