missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM29 Antibody (TRIM29/1042), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47809AF700
This item is not returnable.
View return policy
Description
TRIM29 Monoclonal antibody specifically detects TRIM29 in Human samples. It is validated for Western Blot
Specifications
| TRIM29 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Ataxia telangiectasia group D-associated protein, ATDC, ATDC ataxia-telangiectasia group D-associated protein, FLJ36085, tripartite motif containing 29, tripartite motif protein TRIM29, tripartite motif-containing 29, tripartite motif-containing protein 29 | |
| Recombinant fragment (126 Amino acid residues between aa 1-200) of human TRIM29 protein (Uniprot: Q14134) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| TRIM29/1042 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 23650 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction