missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidylate Synthase Antibody (SPM453), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34441CL1
This item is not returnable.
View return policy
Description
Thymidylate Synthase Monoclonal antibody specifically detects Thymidylate Synthase in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Flow (Intracellular),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Thymidylate Synthase | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422 | |
| Recombinant full-length human Thymidylate Synthase protein. (Uniprot: P04818 ) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control | |
| 7298 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| SPM453 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction