missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFII-I Antibody (PCRP-GTF2I-1A7) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24207JF635
This item is not returnable.
View return policy
Description
TFII-I Monoclonal antibody specifically detects TFII-I in Human samples. It is validated for Flow Cytometry
Specifications
| TFII-I | |
| Monoclonal | |
| Janelia Fluor 635 | |
| BAP-135,135kD, BAP135WBS, Bruton tyrosine kinase-associated protein 135, BTKAP1, DIWS, FLJ38776, FLJ56355, general transcription factor II, i, general transcription factor IIi, general transcription factor II-I, GTFII-I, IB291, SPINBTK-associated protein 135, TFII-ISRF-Phox1-interacting protein, WBSCR6, Williams-Beuren syndrome chromosome region 6 | |
| Recombinant human TFII-I protein | |
| 0.1 mL | |
| Signal Transduction, Transcription Factors and Regulators | |
| 2969 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| PCRP-GTF2I-1A7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction