missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau [p Ser202, p Thr205] Antibody (AH36) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18265JF585
This item is not returnable.
View return policy
Description
Tau Monoclonal antibody specifically detects Tau in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Dot Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Tau | |
| Monoclonal | |
| Janelia Fluor 585 | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Synthetic peptide of Human Phospho Tau (Ser202/Thr205) | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| 4137 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Western Blot, Dot Blot, Immunocytochemistry/Immunofluorescence | |
| AH36 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction