missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPBPR Antibody (1059329), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11337PECY7
This item is not returnable.
View return policy
Description
TAPBPR Monoclonal antibody specifically detects TAPBPR in Human samples. It is validated for Flow Cytometry
Specifications
| TAPBPR | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55080 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 1059329 | |
| Flow Cytometry | |
| FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBP-RTAP binding protein related, TAPBPRtapasin-like | |
| Chinese Hamster Ovary cell line, CHO-derived human TAPBPR, Ala19-Arg404, Accession # Q9BX59.2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction