missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF9 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41266UV
This item is not returnable.
View return policy
Description
TAF9 Polyclonal antibody specifically detects TAF9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TAF9 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| EC 2.7.4.3, MGC:5067, MGC1603, RNA polymerase II TBP-associated factor subunit G, STAF31/32, TAF2G, TAF9, TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa, TAFII31, TAFII-31, TAFII32, TAFII-32, TAFIID32, transcription initiation factor TFIID 31 kD subunit, Transcription initiation factor TFIID 31 kDa subunit, Transcription initiation factor TFIID 32 kDa subunit, transcription initiation factor TFIID subunit 9 | |
| Antibody was raised against a 17 amino acid peptide near the center of human TAF9. The immunogen is located within amino acids 110 - 160 of TAF9. Amino Acid Squence: LTAPNYRLKSLQKKAS | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6880 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction