missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Symplekin Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74590AF750
This item is not returnable.
View return policy
Description
Symplekin Polyclonal antibody specifically detects Symplekin in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Symplekin | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| FLJ27092, SPKSYM, symplekin | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Symplekin using the numbering given in entry NP_004810.2 (GeneID 8189). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8189 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction