missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP-D Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90939CL1
This item is not returnable.
View return policy
Description
SP-D Monoclonal antibody specifically detects SP-D in Macaque samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SP-D | |
| Monoclonal | |
| CoraFluor 1 | |
| COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus SP-D (Accession#: NP_001035283.1; Met1-Phe375). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6441 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction