missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin Antibody (SST/7200R) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20975JF585
This item is not returnable.
View return policy
Description
Somatostatin Monoclonal antibody specifically detects Somatostatin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Somatostatin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 | |
| Recombinant fragment of human Somatostatin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 6750 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SST/7200R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction