missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC44A4 Antibody (ASG-5ME) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28624JF669
This item is not returnable.
View return policy
Description
SLC44A4 Monoclonal antibody specifically detects SLC44A4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SLC44A4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| C6orf29, Choline Transporter-Like Protein 4, Chromosome 6 Open Reading Frame 29, CTL4, NG22, Solute Carrier Family 44 Member 4, Solute Carrier Family 44, Member 4 | |
| SLC44A4 | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 80736 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| ASG-5ME | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction