missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A5 Antibody (idactamab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28813JF525
This item is not returnable.
View return policy
Description
SLC1A5 Monoclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SLC1A5 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| AAAT, Alanine/serine/cysteine transporter 2, ASC amino acid transporter-2, ASCT2, ATB(0), ATBO, Baboon M7 virus receptor, M7V1, M7VS1, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
| SLC1A5/ ASCT2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| idactamab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 6510 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction