missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP gamma/CD172g Antibody (KWAR23) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28671JF525
This item is not returnable.
View return policy
Description
SIRP gamma/CD172g Monoclonal antibody specifically detects SIRP gamma/CD172g in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SIRP gamma/CD172g | |
| Monoclonal | |
| Janelia Fluor 525 | |
| bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma | |
| SIRPg/ CD172g | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55423 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| KWAR23 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction