missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SH2D1A Antibody (13) [Allophycocyanin], Novus Biologicals™
Shop All Bio Techne ProductsDescription
SH2D1A Monoclonal antibody specifically detects SH2D1A in Human samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
Specifications
| Antigen | SH2D1A |
| Applications | ELISA, Sandwich ELISA Detection |
| Classification | Monoclonal |
| Clone | 13 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SH2D1A (RefSeq#: NP_002342.1; Pro2-Lys97). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?