missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Serpin B5/Maspin Antibody (SERPINB5/4974) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26988AF700
This item is not returnable.
View return policy
Description
Serpin B5/Maspin Monoclonal antibody specifically detects Serpin B5/Maspin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Serpin B5/Maspin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
| Recombinant fragment of human Serpin B5/Maspin protein (around aa1-200) (exact sequence is proprietary) | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Tumor Suppressors | |
| 5268 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| SERPINB5/4974 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction