missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 3A Antibody (215803) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB1250X

Product Code. 18563425

  • £548.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Semaphorin 3A Monoclonal antibody specifically detects Semaphorin 3A in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Semaphorin 3A
Monoclonal
Alexa Fluor 532
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
215803
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
(semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3A, Lys26-Val771 (Arg555Ala, Arg552Ala), Accession # Q14563
0.1 mL
Neuroscience
10371
Store at 4°C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.