missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-arrestin Antibody (S128), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25161JF549
This item is not returnable.
View return policy
Description
S-arrestin Monoclonal antibody specifically detects S-arrestin in Human, Mouse, Rat, Porcine, Bovine, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| S-arrestin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin | |
| Recombinant bovine S-arrestin with the first 20 amino acids of the C-terminus truncated [UniProt# P08168] | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 6295 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| S128 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction