missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1T1/ETO Antibody (OTI1H1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73956CL1
This item is not returnable.
View return policy
Description
RUNX1T1/ETO Monoclonal antibody specifically detects RUNX1T1/ETO in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RUNX1T1/ETO | |
| Monoclonal | |
| CoraFluor 1 | |
| AML1T1protein CBFA2T1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, Protein ETO, Protein MTG8, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22 | |
| Full length human recombinant protein of human RUNX1T1(NP_783553) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1H1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 862 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction